missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 98+%

Manufacturer:  Alfa Aesar™ H5469314

 View more versions of this product

Catalog No. AAH5469314



Specifications

Specifications

Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate
145°C
133894-48-1
MFCD00210035
6842341
BSCYRXJVGSZNKX-UHFFFAOYSA-N
chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
16211753
≥98%
Moisture sensitive
25g
C12H24ClF6N3P2
UN1759
acmc-1c01w
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F
421.733
421.73

Chemical Identifiers

133894-48-1
421.733
BSCYRXJVGSZNKX-UHFFFAOYSA-N
16211753
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F
C12H24ClF6N3P2
MFCD00210035
acmc-1c01w
chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

TSCA : No

Recommended Storage : Keep cold; Store under Nitrogen

Documents