missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, 98+%

Manufacturer:  Alfa Aesar™ H5469303

 View more versions of this product

Catalog No. AAH5469303



Specifications

Specifications

Chlorotri(1-pyrrolidinyl)phosphonium hexafluorophosphate
1g
C12H24ClF6N3P2
UN1759
acmc-1c01w
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F
421.733
421.73
145°C
133894-48-1
MFCD00210035
6842341
BSCYRXJVGSZNKX-UHFFFAOYSA-N
chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
16211753
≥98%

Chemical Identifiers

133894-48-1
421.733
BSCYRXJVGSZNKX-UHFFFAOYSA-N
16211753
C1CCN(C1)[P+](N2CCCC2)(N3CCCC3)Cl.F[P-](F)(F)(F)(F)F
C12H24ClF6N3P2
MFCD00210035
acmc-1c01w
chloro(tripyrrolidin-1-yl)phosphanium;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

Recommended Storage : 2° to 8°C

Documents