Please login to your online account to display your discounted pricing

Chlorotitanium triisopropoxide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Chlorotitanium triisopropoxide
Name Note 98%
CAS 20717-86-6
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C9H21ClO3Ti
Linear Formula [(CH3)2CHO]3TiCl
Formula Weight 260.61
MDL Number MFCD00009861
Synonym Chlorotriisopropyloxytitane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC224590010

  SDS
thermo fisher scientific
AC224590010
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid or Low Melting Solid, or Crystals
Assay ≥97.5%
Formula Weight 260.61
Quantity 1kg
Color Colorless to yellow
Density 1.0900g/mL
Name Note 98%
Assay Percent Range 98%
Molecular Formula C9H21ClO3Ti
Linear Formula [(CH3)2CHO]3TiCl
MDL Number MFCD00009861
CAS 20717-86-6
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Infrared Spectrum Authentic
Chemical Name or Material Chlorotitanium triisopropoxide
Synonym Chlorotriisopropyloxytitane
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.