missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorotitanium triisopropoxide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC224590010

 View more versions of this product

Catalog No. AC224590010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Chlorotitanium triisopropoxide
Colorless to yellow
98%
Authentic
≥97.5%
C9H21ClO3Ti
260.61
Chlorotriisopropyloxytitane
1.0900g/mL
1kg
20717-86-6
Liquid or Low Melting Solid, or Crystals
98%
[(CH3)2CHO]3TiCl
MFCD00009861

Chemical Identifiers

20717-86-6
C9H21ClO3Ti
260.61
Chlorotriisopropyloxytitane
≥97.5%
98%
[(CH3)2CHO]3TiCl
MFCD00009861
Chlorotitanium triisopropoxide, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger