Please login to your online account to display your discounted pricing

chlorosulfonylacetyl chloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material chlorosulfonylacetyl chloride
Assay ≥97.5% (Argentometry)
Assay Percent Range 98%
Molecular Formula C2H2Cl2O3S
Linear Formula ClSOCHCOCl
Formula Weight 177.01
MDL Number MFCD00007457
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC369210050

  SDS
thermo fisher scientific
AC369210050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC369210010

  SDS
thermo fisher scientific
AC369210010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (Argentometry)
Physical Form Powder, Crystals and/or Chunks
Formula Weight 177.01
Quantity 5g
Color Off-white to brown
Density 1.669g/mL
UN Number 3265
Linear Formula ClSOCHCOCl
Assay Percent Range 98%
CAS 4025-77-8
Chemical Name or Material chlorosulfonylacetyl chloride
Molecular Formula C2H2Cl2O3S
MDL Number MFCD00007457
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
Contact with water liberates toxic gas.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Alkylation reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently with water.
Contact with water liberates toxic gas.