missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorosulfonyl isocyanate, 98+%, ACROS Organics™

Manufacturer:  Acros Organics AC162040025

 View more versions of this product

Catalog No. AC162040025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Alkylation reagent, C-X bond formation reagent
Specifications

Specifications

Chloropyrosulfonyl isocyanate: max. 1.0%
1.6260g/mL
106.0°C
-44.0°C
1189-71-5
Glass bottle
≥98%
CClNO3S
01,117; 02,70; 03,51; 04,90; 05,132; 06,122; 07,65; 08,105; 10,94; 11,125; 12,122; 13,80
WRJWRGBVPUUDLA-UHFFFAOYSA-N
N-(oxomethylidene)sulfamoyl chloride
70918
Crystals, Crystalline Powder or Granules
Chlorosulfonyl isocyanate
>110°C
White
2.5L
Authentic
1.4450 to 1.4490
98+%
ClSO2NCO
CSI
C(=NS(=O)(=O)Cl)=O
141.525
141.53

Chemical Identifiers

1189-71-5
141.525
CSI
CClNO3S
WRJWRGBVPUUDLA-UHFFFAOYSA-N
70918
SDS Documents