missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroquine Diphosphate Salt MP Biomedicals

$48.25 - $48.25

Chemical Identifiers

CAS 50-63-5
Molecular Formula C18H26ClN3·2H3O4P
Formula Weight 515.9
Synonym Chloroquinone Phosphate
Chemical Name or Material Chloroquine Diphosphate Salt
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19391910 Mp Biomedicals
19391910
10g Each for $48.25
Specifications Description & Specifications

Chemical Identifiers

50-63-5
515.9
Chloroquine Diphosphate Salt
C18H26ClN3·2H3O4P
Chloroquinone Phosphate

Specifications

515.9
Chloroquinone Phosphate
50-63-5
10g
C18H26ClN3·2H3O4P
Chloroquine Diphosphate Salt