Please login to your online account to display your discounted pricing

Chloroplatinic Acid Hexahydrate (Certified ACS), Fisher Chemical

$1,058.15 - $1,058.15

Chemical Identifiers

Chemical Name or Material Chloroplatinic Acid Hexahydrate
CAS 18497-13-7
CAS 16941-12-1
CAS 26023-84-7
Purity Grade Certified ACS Grade
Assay ≥37.50%
Molecular Formula H2Cl6Pt · x H2O
Formula Weight 517.91
MDL Number MFCD00149910
Synonym Dihydrogen Hexachloroplatinum (IV) Hexahydrate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

P154-1

  SDS
fisher scientific company
P1541
Glass Ampoule 1g Each for $1,058.15
Case of 6 Each for $6,348.90
Description & Specifications

Specifications

Assay ≥37.50%
Purity Grade Certified ACS Grade
Physical Form Solid Powder
Formula Weight 517.91
Melting Point 60°C
Quantity 1g
Packaging Glass Ampoule
UN Number UN 2507
Density 2.43g/mL
DOT Information DOT Class 8, : Corrosive
Molecular Formula H2Cl6Pt · x H2O
Color Amber
Solubility Pass Test (in ethanol)
Health Hazard 3 Emergency Overview
Toxic if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction. May cause sensitization by inhalation and skin contact. Water reactive. Contact with water liberates toxic gas.
NFPA
Health:3 Flammability:0 Instability:1
CAS 26023-84-7
CAS 18497-13-7
Chemical Name or Material Chloroplatinic Acid Hexahydrate
Identification Pass Test
Alkalis and Other Salts 0.05% max.
Suitability Pass Test
MDL Number MFCD00149910
Synonym Dihydrogen Hexachloroplatinum (IV) Hexahydrate
Health Hazard 1 Signal Word:DANGER!
CAS 16941-12-1
ChemAlert Storage Symbol White

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Signal Word:DANGER!

Emergency Overview
Toxic if swallowed. Causes burns by all exposure routes. May cause allergic respiratory and skin reaction. May cause sensitization by inhalation and skin contact. Water reactive. Contact with water liberates toxic gas.
NFPA
Health:3 Flammability:0 Instability:1