missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chlorophyll (Total Chlorophyll: ca. 6.0%), TCI America™

Manufacturer:  Tci America C078025G

Catalog No. C078025G



Specifications

Specifications

Chlorophyll (Total Chlorophyll: ca. 6.0%)
25g
C55H72MgN4O5
2s-1-4r,7s,10s,13s,16s,19r-19-amino-7-2-amino-2-oxo-ethyl-10-3-amino-3-oxo-propyl-13-benzyl-16-4-hydroxyphenyl methyl-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl-n-1s-1-2-amino-2-oxo-ethyl carbamoyl-4-guanidino-butyl pyrro, 2s-1-4r,7s,10s,13s,16s,19r-19-amino-7-2-amino-2-oxo-ethyl-10-3-amino-3-oxo-propyl-13-benzyl-16-4-hydroxyphenyl methyl-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl-n-1s-5-amino-1-2-amino-2-oxo-ethyl carbamoyl pentyl pyrrolid, chlorophyll, chlorophyll a, chlorophyll-a, magnesium, 2e,7r,11r-3,7,11,15-tetramethyl-2-hexadecenyl 3s,4s,21r-9-ethenyl-14-ethyl-21-methoxycarbonyl-4,8,13,18-tetramethyl-20-oxo-3-phorbinepropanoato 2--kappan23,kappan24,kappan25,kappan26-, sp-4-2, magnesium, 3,7,11,15-tetramethyl-2-hexadecenyl 9-ethenyl-14-ethyl-21-methoxycarbonyl-4,8,13,18-tetramethyl-20-oxo-3-phorbinepropanoato 2--n23,n24,n25,n26-, sp-4-2-3s-3alpha 2e,7s*,11s* ,4beta,21beta, unii-00wnz48or9 component, unii-yf5q9ejc8y, yf5q9ejc8y
CCC1=C(C2=NC1=CC3=C(C4=C(C(C(=C4[N-]3)C5=NC(=CC6=NC(=C2)C(=C6C)C=C)C(C5CCC(=O)OCC=C(C)CCCC(C)CCCC(C)CCCC(C)C)C)C(=O)OC)[O-])C)C.[Mg+2]
16667503
Crystal-Powder at 20°C
Undesignated
1406-65-1
MFCD00071051
ATNHDLDRLWWWCB-AENOIHSZSA-M
893.509
CHEBI:18230

Chemical Identifiers

1406-65-1
893.509
ATNHDLDRLWWWCB-AENOIHSZSA-M
16667503
CCC1=C(C2=NC1=CC3=C(C4=C(C(C(=C4[N-]3)C5=NC(=CC6=NC(=C2)C(=C6C)C=C)C(C5CCC(=O)OCC=C(C)CCCC(C)CCCC(C)CCCC(C)C)C)C(=O)OC)[O-])C)C.[Mg+2]
C55H72MgN4O5
MFCD00071051
2s-1-4r,7s,10s,13s,16s,19r-19-amino-7-2-amino-2-oxo-ethyl-10-3-amino-3-oxo-propyl-13-benzyl-16-4-hydroxyphenyl methyl-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl-n-1s-1-2-amino-2-oxo-ethyl carbamoyl-4-guanidino-butyl pyrro, 2s-1-4r,7s,10s,13s,16s,19r-19-amino-7-2-amino-2-oxo-ethyl-10-3-amino-3-oxo-propyl-13-benzyl-16-4-hydroxyphenyl methyl-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl-n-1s-5-amino-1-2-amino-2-oxo-ethyl carbamoyl pentyl pyrrolid, chlorophyll, chlorophyll a, chlorophyll-a, magnesium, 2e,7r,11r-3,7,11,15-tetramethyl-2-hexadecenyl 3s,4s,21r-9-ethenyl-14-ethyl-21-methoxycarbonyl-4,8,13,18-tetramethyl-20-oxo-3-phorbinepropanoato 2--kappan23,kappan24,kappan25,kappan26-, sp-4-2, magnesium, 3,7,11,15-tetramethyl-2-hexadecenyl 9-ethenyl-14-ethyl-21-methoxycarbonyl-4,8,13,18-tetramethyl-20-oxo-3-phorbinepropanoato 2--n23,n24,n25,n26-, sp-4-2-3s-3alpha 2e,7s*,11s* ,4beta,21beta, unii-00wnz48or9 component, unii-yf5q9ejc8y, yf5q9ejc8y
CHEBI:18230
Safety and Handling

Safety and Handling

RTECSNumber : FW6420000

TSCA : No

Documents