Please login to your online account to display your discounted pricing

Chlorophenol Red Sodium Salt, Reagent Grade, LabChem™

$189.69 - $189.69

Chemical Identifiers

Chemical Name or Material Chlorophenol Red Sodium Salt
CAS 123333-64-2
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C19H11Cl2O5SNa
Formula Weight 445.25
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC130707

 
lab chem
LC130707
10g Each for $189.69
Description & Specifications

Specifications

Formula Weight 445.25
Identification Passes Test
Purity Grade Reagent Grade
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
Color Maroon
Quantity 10g
Linear Formula C19H11Cl2O5SNa
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.
Health Hazard 3 GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Keep container tightly closed.
Store locked up in a well-ventilated place.
Chemical Name or Material Chlorophenol Red Sodium Salt
CAS 123333-64-2
Physical Form Crystals
CAS Max % 100
Solubility Soluble in water
Recommended Storage Room Temperature
Packaging Amber Glass
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
Avoid breathing dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Use only outdoors or in a well-ventilated area.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Keep container tightly closed.
Store locked up in a well-ventilated place.

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.

Room Temperature