Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorophenol Red sodium salt

$53.73 - $53.73

Chemical Identifiers

Chemical Name or Material Chlorophenol Red sodium salt
CAS 123333-64-2
Molecular Formula C19H11Cl2NaO5S
Formula Weight 445.26
MDL Number MFCD00151199
Synonym Chlorophenol Red, water soluble
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1036406

 
alfa aesar™
L1036406
5g Each for $53.73
Description & Specifications

Specifications

Formula Weight 445.26
Chemical Name or Material Chlorophenol Red sodium salt
Molecular Formula C19H11Cl2NaO5S
MDL Number MFCD00151199
CAS 123333-64-2
Synonym Chlorophenol Red, water soluble
Melting Point ≈267°C
Quantity 5g

Applications
Chlorophenol Red sodium salt is used in the standard samples to indicate pH. A stain used to detect chlorine dioxide in drinking water.

Solubility
Soluble in water.

Notes
Transition interval: pH 4.8 (yellow) to 6.4 (red)