Please login to your online account to display your discounted pricing

Alfa Aesar™ Chlorophenol Red sodium salt, 0.04% w/v aq. soln.

Chemical Identifiers

Chemical Name or Material Chlorophenol Red sodium salt
Formula Weight 445.26
MDL Number MFCD00151199
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38699AE

 
alfa aesar™
38699AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38699AP

 
alfa aesar™
38699AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38699M0

 
alfa aesar™
38699M0
6 x 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 445.26
Chemical Name or Material Chlorophenol Red sodium salt
Quantity 100mL
TSCA TSCA
Concentration 0.04% w/v aq. soln.
Physical Form Liquid
MDL Number MFCD00151199
Density 0.990g/mL