Please login to your online account to display your discounted pricing

Chlorophenol Red Indicator, 0.04% (w/v) Aqueous Solution, Ricca Chemical


Quantity 250mL

Manufacturer: ricca chemical company  RIR2000000250A

 View more versions of this product

Catalog No. R2000000250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Chlorophenol Red Indicator
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 123333-64-2
CAS Min % 0.04
CAS Max % 0.04
Physical Form ear, red liquid
Quantity 250mL

Specifications

Chemical Name or Material Chlorophenol Red Indicator
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 123333-64-2
CAS Min % 0.04
CAS Max % 0.04
Physical Form ear, red liquid