Please login to your online account to display your discounted pricing

Chloropentafluorobenzene, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Chloropentafluorobenzene
Name Note 99%
CAS 344-07-0
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C6ClF5
Linear Formula ClC6F5
Formula Weight 202.51g/mol
MDL Number MFCD00000534
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC186070050

  SDS
fisher scientific acros
186070050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4220 to 1.4240 (20°C, 589nm)
Color Clear colorless
Quantity 5g
Chemical Name or Material Chloropentafluorobenzene
Name Note 99%
CAS 344-07-0
Physical Form Clear colorless liquid
Assay 98.5% min.
Assay Percent Range 99%
Molecular Formula C6ClF5
Linear Formula ClC6F5
Formula Weight 202.51g/mol
MDL Number MFCD00000534
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/eye protection/face protection.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number CZ1225500
EINECS Number 206-450-6
TSCA TSCA
Safety and Handling

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/eye protection/face protection.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.