Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloropentacarbonylrhenium(I), 98%

Chemical Identifiers

Chemical Name or Material Chloropentacarbonylrhenium(I)
CAS 14099-01-5
Linear Formula Re(CO)5Cl
Formula Weight 361.71
MDL Number MFCD00013296
UN Number UN3466
Synonym Rhenium(I) pentacarbonyl chloride; Pentacarbonylchlororhenium(I)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4209603

  SDS
alfa aesar™
4209603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 361.71
Chemical Name or Material Chloropentacarbonylrhenium(I)
Physical Form Crystalline
Assay 98%
MDL Number MFCD00013296
Synonym Rhenium(I) pentacarbonyl chloride; Pentacarbonylchlororhenium(I)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: METAL CARBONYLS, SOLID, N.O.S.
Linear Formula Re(CO)5Cl
CAS 14099-01-5
UN Number UN3466
Color White to pale brown
Quantity 1g
Health Hazard 3 GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Sensitivity Air & Moisture Sensitive
Safety and Handling

Danger

GHS P Statement
P280f-P309-P310
Wear protective clothing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.