Learn More
Chloropentaamminecobalt(III) chloride, Thermo Scientific Chemicals
$101.34 - $918.78
Chemical Identifiers
CAS | 13859-51-3 |
---|---|
MDL Number | MFCD00064728 |
Synonym | chlorpentaaminecobalt, chloropentaamminecobalt iii dichloride, pentaaminechlorocobalt dichloride, pentaamminechlorocobalt dichloride, chloropentaminecobalt iii chloride, pentaaminechlorocobalt iii chloride, cobalt 3+ chloropentammine chloride, pentaamminechlorocobalt 2+ dichloride, cobalt 3+ , pentaammine-, trichloride, cobalt 2+ , pentaamminechloro-, dichloride, oc-6-22 |
Description
Chloropentaamminecobalt(III) chloride is doped with polyvinyl acetate to prepare polymeric films, which enhances the optical conductivity. It is also used to prepare the corresponding hydrogen sulfate complex by reacting with sulfuric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13859-51-3 | |
MFCD00064728 | |
chlorpentaaminecobalt, chloropentaamminecobalt iii dichloride, pentaaminechlorocobalt dichloride, pentaamminechlorocobalt dichloride, chloropentaminecobalt iii chloride, pentaaminechlorocobalt iii chloride, cobalt 3+ chloropentammine chloride, pentaamminechlorocobalt 2+ dichloride, cobalt 3+ , pentaammine-, trichloride, cobalt 2+ , pentaamminechloro-, dichloride, oc-6-22 |
Specifications
13859-51-3 | |
Purple | |
Odorless | |
MFCD00064728 | |
chlorpentaaminecobalt, chloropentaamminecobalt iii dichloride, pentaaminechlorocobalt dichloride, pentaamminechlorocobalt dichloride, chloropentaminecobalt iii chloride, pentaaminechlorocobalt iii chloride, cobalt 3+ chloropentammine chloride, pentaamminechlorocobalt 2+ dichloride, cobalt 3+ , pentaammine-, trichloride, cobalt 2+ , pentaamminechloro-, dichloride, oc-6-22 | |
250.44 | |
Chloropentaamminecobalt(III) chloride |
(decomposition) | |
1.819 g/mL | |
[CoCl(NH3)5]Cl2 | |
2 g | |
Soluble in water. | |
Powder |
Safety and Handling
GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H350
EINECSNumber : 237-594-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only