missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Chloropentaamminecobalt(III) chloride

Manufacturer:  Alfa Aesar™ 1376518

 View more versions of this product

Catalog No. AA1376518



Specifications

Specifications

Chloropentaamminecobalt(III) chloride
Odorless
50g
[CoCl(NH3)5]Cl2
chlorpentaaminecobalt, chloropentaamminecobalt iii dichloride, pentaaminechlorocobalt dichloride, pentaamminechlorocobalt dichloride, chloropentaminecobalt iii chloride, pentaaminechlorocobalt iii chloride, cobalt 3+ chloropentammine chloride, pentaamminechlorocobalt 2+ dichloride, cobalt 3+ , pentaammine-, trichloride, cobalt 2+ , pentaamminechloro-, dichloride, oc-6-22
250.44
1.819g/mL
Purple
13859-51-3
MFCD00064728
Slightly soluble in water, soluble in conc; H2SO4; Insoluble in alcohol
Powder

Chemical Identifiers

13859-51-3
MFCD00064728
chlorpentaaminecobalt, chloropentaamminecobalt iii dichloride, pentaaminechlorocobalt dichloride, pentaamminechlorocobalt dichloride, chloropentaminecobalt iii chloride, pentaaminechlorocobalt iii chloride, cobalt 3+ chloropentammine chloride, pentaamminechlorocobalt 2+ dichloride, cobalt 3+ , pentaammine-, trichloride, cobalt 2+ , pentaamminechloro-, dichloride, oc-6-22
Safety and Handling

Safety and Handling

GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.

GHS P Statement
P280h-P262
Wear protective gloves/clothing.
Do not get in eyes, on skin, or on clothing.

Warning

TSCA : TSCA

Documents