missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloromethyltrimethylsilane, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 194761000

 View more versions of this product

Catalog No. AC194761000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

Chloromethyltrimethylsilane
2344-80-1
0.8790g/mL
−2°C
98+%
C4H11ClSi
(CH3)3SiCH2Cl
100 g
12,118; 13,76
chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane
OOCUOKHIVGWCTJ-UHFFFAOYSA-N
chloromethyl(trimethyl)silane
75361
98+%
98+%
Colorless
97.0°C to 98.0°C
Authentic
Glass bottle
1.4165 to 1.4185
MFCD00000878
04, III, 1844
0.879
Solubility in water: insoluble
C[Si](C)(C)CCl
122.67
122.67
Liquid

Chemical Identifiers

2344-80-1
122.67
OOCUOKHIVGWCTJ-UHFFFAOYSA-N
75361
C[Si](C)(C)CCl
C4H11ClSi
MFCD00000878
chloromethyl trimethylsilane, silane, chloromethyl trimethyl, trimethylsilyl methyl chloride, chloromethyl trimethyl silane, ch3 3sich2cl, tmcms, pubchem13668, trimethylchloromethylsilane
chloromethyl(trimethyl)silane
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-058-5

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only