missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloromethyl pivalate, 99+%, Acros Organics

Manufacturer:  Thermo Fisher Scientific AC161465000

 View more versions of this product

Catalog No. AC161465000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Chloromethyl pivalate
40°C
Glass bottle
1.038
Colorless to Yellow
18997-19-8
C6H11ClO2
MFCD00000884
Solubility in water: insoluble. Other solubilities: soluble in alcohol,toluene and chlorinated,hydrocarbons
CC(C)(C)C(=O)OCCl
150.6
150.6
99+%
1.0380g/mL
Authentic
1.4160 to 1.418
146°C to 148°C
500g
99% min. (GC)
(CH3)3CCO2CH2Cl
2,2-dimethylpropionic acid chloromethyl ester
GGRHYQCXXYLUTL-UHFFFAOYSA-N
chloromethyl 2,2-dimethylpropanoate
87885
Liquid

Chemical Identifiers

18997-19-8
150.6
GGRHYQCXXYLUTL-UHFFFAOYSA-N
87885
CC(C)(C)C(=O)OCCl
C6H11ClO2
MFCD00000884
2,2-dimethylpropionic acid chloromethyl ester
chloromethyl 2,2-dimethylpropanoate
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 242-735-1

SDS Documents