missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloromethyl phenyl sulfone, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 428740050

 View more versions of this product

Catalog No. AC428740050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Chloromethyl phenyl sulfone
113°C
Glass bottle
C6H5SO2CH2Cl
5g
NXAIQSVCXQZNRY-UHFFFAOYSA-N
chloromethylsulfonylbenzene
81625
99%
7205-98-3
98.5% min. (GC)
C7H7ClO2S
MFCD00007551
chloromethyl phenyl sulfone, chloromethyl sulfonyl benzene, phenyl chloromethyl sulfone, chloromethylphenylsulfone, benzene, chloromethyl sulfonyl, chloromethylphenyl sulfone, 1-chloromethylsulfonyl benzene, chloromethyl sulphonyl benzene
C1=CC=C(C=C1)S(=O)(=O)CCl
190.65
190.65

Chemical Identifiers

7205-98-3
190.65
NXAIQSVCXQZNRY-UHFFFAOYSA-N
81625
C1=CC=C(C=C1)S(=O)(=O)CCl
C7H7ClO2S
MFCD00007551
chloromethyl phenyl sulfone, chloromethyl sulfonyl benzene, phenyl chloromethyl sulfone, chloromethylphenylsulfone, benzene, chloromethyl sulfonyl, chloromethylphenyl sulfone, 1-chloromethylsulfonyl benzene, chloromethyl sulphonyl benzene
chloromethylsulfonylbenzene
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 230-581-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only