Learn More
(Chloromethylene)dimethylammonium chloride, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2417214
Description
(Chloromethylene)dimethylammonium chloride is used in the synthesis of beta-lactams, n-vinyl substituted indoles and fused 2-pyridone rings and 2-azetidinones. It is used as a formylating agent and used in the preparation of anti-poxvirus agents. It is used in Staudinger reaction and Hundsdiecker-barton iododecarboxylation reaction. It is widely used for studying organic photosensitizers, which finds application in dye-sensitized solar cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(Chloromethylene)dimethylammonium chloride is used in the synthesis of beta-lactams, n-vinyl substituted indoles and fused 2-pyridone rings and 2-azetidinones. It is used as a formylating agent and used in the preparation of anti-poxvirus agents. It is used in Staudinger reaction and Hundsdiecker-barton iododecarboxylation reaction. It is widely used for studying organic photosensitizers, which finds application in dye-sensitized solar cells.
Solubility
Soluble in water.
Notes
Moisture sensitive. It may decompose in moist air/ water. Incompatible with bases, strong oxidizing agents, amines, metals and butyl rubber.
Specifications
(Chloromethylene)dimethylammonium Chloride | |
130°C (decomposition) | |
C3H7Cl2N | |
25 g | |
3566322 | |
arnold's reagent, vilsmeier reagent, chloromethylene dimethyliminium chloride, methanaminium, n-chloromethylene-n-methyl-, chloride, chloromethylene dimethylammonium chloride, n-chloromethylidene-n-methylmethanaminium chloride, chloromethylenedimethylammonium chloride, chloromethylidene dimethylazanium chloride, n-chloromethylene-n,n-dimethyl ammonium chloride | |
QQVDYSUDFZZPSU-UHFFFAOYSA-M | |
chloromethylidene(dimethyl)azanium;chloride | |
77311 | |
96% |
3724-43-4 | |
Acrid | |
MFCD00011868 | |
3261 | |
Moisture sensitive | |
Soluble in water. | |
C[N+](=CCl)C.[Cl-] | |
127.996 | |
128 |
Chemical Identifiers
3724-43-4 | |
MFCD00011868 | |
77311 |
C3H7Cl2N | |
QQVDYSUDFZZPSU-UHFFFAOYSA-M | |
chloromethylidene(dimethyl)azanium;chloride |
Safety and Handling
GHS H Statement
H360-H314-H318-H302
May damage fertility or the unborn child.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P201-P202-P234-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c
H290-H302-H314-H335-H360D
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 425-970-6
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only