Please login to your online account to display your discounted pricing

(Chloromethyl)triphenylphosphonium chloride, 95%, tech., ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  109530250

 View more versions of this product

Catalog No. AC109530250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Melting Point 258° to 264°C (dec.)
Physical Form Fine Crystalline Powder
Purity Grade Technical Grade
Assay ≥94%
Formula Weight 347.21
Quantity 25g
Color White to almost white
RTECS Number TA1882000
EINECS Number 226-139-9
TSCA TSCA
Fieser 07,62
CAS Max % 100.0
CAS Min % 94.0
MDL Number MFCD00011867
Molecular Formula C19H17Cl2P
Chemical Name or Material (Chloromethyl)triphenylphosphonium chloride
Name Note tech., 95%
Assay Percent Range 95%
Linear Formula ClCH2P(C6H5)3Cl
CAS 5293-84-5
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material (Chloromethyl)triphenylphosphonium chloride
Name Note tech., 95%
CAS 5293-84-5
CAS Min % 94.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C19H17Cl2P
Linear Formula ClCH2P(C6H5)3Cl
Formula Weight 347.21
MDL Number MFCD00011867
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents