Please login to your online account to display your discounted pricing

(Chloromethyl)triphenylphosphonium chloride, 95%, tech., ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  109530050

 View more versions of this product

Catalog No. AC109530050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Melting Point 258° to 264°C (dec.)
Purity Grade Technical Grade
Physical Form Fine Crystalline Powder
Formula Weight 347.21
Assay ≥94%
Quantity 5g
Color White to almost white
Fieser 07,62
EINECS Number 226-139-9
TSCA TSCA
RTECS Number TA1882000
CAS Min % 94.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 5293-84-5
MDL Number MFCD00011867
Chemical Name or Material (Chloromethyl)triphenylphosphonium chloride
Name Note tech., 95%
Assay Percent Range 95%
Molecular Formula C19H17Cl2P
Linear Formula ClCH2P(C6H5)3Cl
Packaging Glass bottle
Health Hazard 1 Warning

Specifications

Chemical Name or Material (Chloromethyl)triphenylphosphonium chloride
Name Note tech., 95%
CAS 5293-84-5
CAS Min % 94.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C19H17Cl2P
Linear Formula ClCH2P(C6H5)3Cl
Formula Weight 347.21
MDL Number MFCD00011867
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents