Please login to your online account to display your discounted pricing

(Chloromethyl)triphenylphosphonium chloride, 95%, tech., ACROS Organics™

Chemical Identifiers

Chemical Name or Material (Chloromethyl)triphenylphosphonium chloride
Name Note tech., 95%
CAS 5293-84-5
CAS Min % 94.0
CAS Max % 100.0
Purity Grade Technical Grade
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C19H17Cl2P
Linear Formula ClCH2P(C6H5)3Cl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC109530250

  SDS
fisher scientific acros
109530250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC109530050

  SDS
thermo fisher scientific
109530050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 258° to 264°C (dec.)
Purity Grade Technical Grade
Physical Form Fine Crystalline Powder
Assay ≥94%
Formula Weight 347.21
Quantity 25g
Color White to almost white
EINECS Number 226-139-9
TSCA TSCA
RTECS Number TA1882000
Fieser 07,62
CAS Max % 100.0
CAS Min % 94.0
Molecular Formula C19H17Cl2P
MDL Number MFCD00011867
Chemical Name or Material (Chloromethyl)triphenylphosphonium chloride
Name Note tech., 95%
Assay Percent Range 95%
Linear Formula ClCH2P(C6H5)3Cl
CAS 5293-84-5
Health Hazard 1 Warning
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Packaging Glass bottle
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For C-C Bond Formation (Olefination)

Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.