Please login to your online account to display your discounted pricing

Alfa Aesar™ (Chloromethyl)trichlorosilane, 97%

Chemical Identifiers

Chemical Name or Material (Chloromethyl)trichlorosilane
CAS 1558-25-4
Molecular Formula CH2Cl4Si
Linear Formula ClCH2SiCl3
Formula Weight 183.93
MDL Number MFCD00000873
UN Number UN3390
Beilstein 1811640
Synonym Trichloro(chloromethyl)silane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1643306

  SDS
alfa aesar™
L1643306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL1643314

  SDS
alfa aesar™
L1643314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 183.93
Assay 97%
Molecular Formula CH2Cl4Si
Chemical Name or Material (Chloromethyl)trichlorosilane
Health Hazard 2 GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
MDL Number MFCD00000873
CAS 1558-25-4
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310-P302+P352-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place.
Odor Acrid
Flash Point 69°C (156°F)
Health Hazard 1 Danger
Boiling Point 117° to 118°C
Synonym Trichloro(chloromethyl)silane
Linear Formula ClCH2SiCl3
Beilstein 1811640
Quantity 5g
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.
UN Number UN3390
RTECS Number VV2200000
TSCA TSCA

Applications
(chloromethyl)trichlorosilane is usede in the synthesis of a highly branched polycarbosilane. Ferrocenophane has been prepared from (chloromethyl)trichlorosilane.

Solubility
Reacts with water.

Notes
Air & moisture sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, air and moisture.

Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P309-P310-P302+P352-P402
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place.

GHS H Statement
H330-H314-H318-H227
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.