missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloromethyl chlorosulfate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 330800010

 View more versions of this product

Catalog No. AC330800010


 

Specifications Description & Specifications

Specifications

≥96.0%
164.99
Clear colorless to yellow
01, III, 2593
Chloromethyl chlorosulfate
97%
Plastic bottle
49715-04-0
Liquid
1g
15, 2147
1.4460 to 1.4480 (20°C, 589nm)
Authentic
MFCD00041547
may pick up color on ageing
CH2Cl2O3S

Chemical Identifiers

49715-04-0
CH2Cl2O3S
MFCD00041547
≥96.0%
97%
164.99
Chloromethyl chlorosulfate

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes severe skin burns and eye damage.
Fatal if inhaled.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger