Please login to your online account to display your discounted pricing

Chloroiodomethane, Stabilized 98%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC222155000

 View more versions of this product

Catalog No. AC222155000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5% (GC)
Formula Weight 176.38
Color Clear pale yellow to pink-reddish
Quantity 500mL
Density 2.42g/mL
Chemical Name or Material Chloroiodomethane
Stabilizer Copper
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula CH2ClI
Linear Formula ClCH2I
CAS 593-71-5
Health Hazard 1 Warning
CAS 7440-22-4
Packaging Glass bottle
Refractive Index 1.5810 to 1.5830 (20°C, 589nm)

Specifications

Chemical Name or Material Chloroiodomethane
CAS 593-71-5
CAS 7440-22-4
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula CH2ClI
Linear Formula ClCH2I
Formula Weight 176.38
Accessories

Warning

SDS & Safety Documents