missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chloroformamidine Hydrochloride 98.0+%, TCI America™
Supplier: TCI America C31995G
Specifications
Chloroformamidine Hydrochloride | |
180°C | |
CH4Cl2N2 | |
5 g | |
C(=N)(N)Cl.Cl | |
114.957 | |
114.96 | |
Crystalline Powder |
29671-92-9 | |
White-Yellow | |
MFCD00035527 | |
FUQFHOLPJJETAP-UHFFFAOYSA-N | |
carbamimidoyl chloride;hydrochloride | |
3084509 | |
≥98.0% (N,T) |
Chemical Identifiers
29671-92-9 | |
114.957 | |
FUQFHOLPJJETAP-UHFFFAOYSA-N | |
carbamimidoyl chloride;hydrochloride |
CH4Cl2N2 | |
MFCD00035527 | |
3084509 | |
C(=N)(N)Cl.Cl |
Safety and Handling
TSCA : No