Learn More
Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals
Chloroform, >-99%, CHCl3, CAS Number-67-66-3, 64-17-5, trichloromethane, trichloroform, trichlormethan, methyl trichloride, cloroformio, methenyl trichloride, formyl trichloride, methane, trichloro, chloroforme, methane trichloride, 2.5L
$145.54 - $257.42
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
SMILES | ClC(Cl)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC390760010
|
Thermo Scientific Chemicals
390760010 |
1 L | Glass Bottle |
Each for $145.54
|
|
||||
AC390760025
|
Thermo Scientific Chemicals
390760025 |
2.5 L | Glass Bottle |
Each for $257.42
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLCChemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
ClC(Cl)Cl |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 |
Specifications
-63.0°C | |
0.009 max. at 300nm, 0.046 max. at 265nm, 0.301 max. at 250nm, 0.699 max. at 245nm, 1 cm cell vs water, 1 cm cell vs water at 0nm | |
1.4800g/mL | |
1 L | |
Crystals | |
0.0002 % max. | |
1.48 | |
CHCl3 | |
0.56 mPa.s (20°C) | |
15, 2142 | |
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton | |
ClC(Cl)Cl | |
119.37 | |
CHEBI:35255 | |
≥99% | |
Chloroform, Stabilized with ethanol |
67-66-3,64-17-5 | |
White | |
61.0°C | |
Glass Bottle | |
1.4443 to 1.4463 | |
99+% | |
CHCl3 | |
MFCD00000826 | |
0.01% Max. (K.F.) | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
trichloromethane | |
6212 | |
119.38 | |
HPLC |
Safety and Handling
GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Warning
RUO – Research Use Only