missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Chloroform, ReagentPlus™, ≥99.8%, Honeywell™

Reagent Grade, contains 0.5-1.0% ethanol as stabilizer,≥99.8%

$142.03 - $142.03

Chemical Identifiers

CAS 67-66-3
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.369
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
IUPAC Name chloroform
SMILES C(Cl)(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
60044790 View Documents Honeywell™
1329501L
1L Glass Bottle Each for $142.03
 
Description

Description

  • ≥95% for general laboratory use.
Specifications

Chemical Identifiers

67-66-3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
chloroform
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
C(Cl)(Cl)Cl

Specifications

67-66-3
CHCl3
UN1888
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
C(Cl)(Cl)Cl
119.369
CHEBI:35255
≥99.8%
1.492g/mL at 25°C, 1.48g/mL at 25°C
n20/D 1.445(lit.)
4.1 (vs air)
60°C to 62°C
-63°C
Chloroform
Contains 0.5 to 1.0% ethanol as stabilizer
MFCD00000826
1731042
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38g/mol
ReagentPlus™
Glass Bottle
160mmHg (20°C)
Liquid
Undesignated
1L
Safety and Handling

Safety and Handling

P260-P280-P301 + P312 + P330-P304 + P340 + P312-P305 + P351 + P338-P403 + P233

H302-H315-H319-H331-H336-H351-H361d-H372

DOTInformation : 6.1/PGIII

EINECSNumber : 200-663-8