Please login to your online account to display your discounted pricing

Chloroform, PHOTREX™ Reagent, J.T.Baker™

High purity solvents characterized for use in UV, visible and IR spectrophotometry

$155.24 - $1,973.75

Chemical Identifiers

Chemical Name or Material Chloroform
CAS 67-66-3
Purity Grade PHOTREX™ Reagent
Molecular Formula CHCl3
Formula Weight 119.38
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002588

 
avantor performance
918303
Narrow Mouth Amber Glass Bottle 4L Each for $1,973.75

02002592

 
avantor performance
918301
Narrow Mouth Amber Glass Bottle 500mL Each for $155.24
Description & Specifications

Specifications

Formula Weight 119.38
Purity Grade PHOTREX™ Reagent
CAS 67-66-3
Chemical Name or Material Chloroform
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.48kg
Quantity 4L
Molecular Formula CHCl3

    For Spectrophotometry