Please login to your online account to display your discounted pricing

Chloroform, OmniPur™, EMD Millipore

$65.47 - $65.47

Chemical Identifiers

Chemical Name or Material Chloroform
CAS 67-66-3
Molecular Formula CHCl3
Formula Weight 119.38
UN Number 1888
Synonym Trichloromethane
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

3150950ML

 
emd millipore corp
3150950ML
Glass Bottle 950mL Each for $65.47
Description & Specifications

Specifications

Chemical Name or Material Chloroform
CAS 67-66-3
Physical Form Liquid
Assay ≥99.8%
Molecular Formula CHCl3
Formula Weight 119.38
UN Number 1888
Synonym Trichloromethane
Solubility Information Partially soluble in water
Odor Pleasant. Ethereal
Packaging Glass Bottle
Refractive Index 1.49
Storage Information Store in accordance with local regulations. Store in original container, protected from direct sunlight. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
Vapor Pressure 22.3 kPa (167mm Hg) [20°C]
Boiling Point 60.5°C
Color Colorless
Melting Point −63°C
Quantity 950mL