missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform, for HPLC, stabilized with amylene, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 268320010

 View more versions of this product

Catalog No. AC268320010



Description

Description

For HPLC
Specifications

Specifications

Chloroform
(1 cm cell vs HPLC−grade water)
(1 cm cell vs HPLC-grade water)
CHCl3
MFCD00000826
01,130; 08,92; 15,84
chloroforme, cloroformio, formyl trichloride, methane trichloride, methane, trichloro, methenyl trichloride, methyl trichloride, trichlormethan, trichloroform, trichloromethane
C(Cl)(Cl)Cl
119.369
CHEBI:35255
Liquid
1.4800g/mL
Glass bottle
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
approx. 100ppm amylene
0.56 mPa.s (20°C)
Undesignated
1L
Stabilized
67-66-3,513-35-9
0.0002 meq/g max.
CHCl3
01, 61
15, 2142
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
HPLC
Yes
0.0005% max.
1.48
0.05% max.
61.0°C
-63.0°C

Chemical Identifiers

67-66-3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
chloroform
CHCl3
MFCD00000826
chloroforme, cloroformio, formyl trichloride, methane trichloride, methane, trichloro, methenyl trichloride, methyl trichloride, trichlormethan, trichloroform, trichloromethane
CHEBI:35255
C(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

EINECSNumber : 200-663-8

RTECSNumber : FS9100000

TSCA : TSCA

SDS Documents