missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform, HPLC Grade, Alfa Aesar™

Manufacturer:  Alfa Aesar 43685M6

 View more versions of this product

Catalog No. AA43685M6



Specifications

Specifications

Chloroform
>99.5%
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
HPLC
Characteristic
Colorless
67-66-3
CHCl3
chloroforme
C(Cl)(Cl)Cl
119.369
CHEBI:35255
Liquid
982°C (1800°F)
158mm Hg at 20°C
4 x 4L

Chemical Identifiers

67-66-3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
chloroform
CHCl3
MFCD00000826
chloroforme
CHEBI:35255
C(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

H351-H373-H302-H315: Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Causes skin irritation.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 200-663-8

Documents