missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform, HPLC Grade, 99.5+% min, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 022920M1

 View more versions of this product

Catalog No. AA22920M1



Description

Description

Chloroform is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Chloroform
67-66-3
Colorless liquid
60.5°C to 61.5°C
Characteristic
1.446
CHCl3
210 hPa (158mm Hg) at 20°C
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
HEDRZPFGACZZDS-UHFFFAOYSA-N
chloroform
6212
119.38
°C to 63°C
Colorless
1.492
4 x 1 L
Liquid
>99.5%
MFCD00000826
1731042
Miscible with diethyl ether,oils,ligroin,alcohol,carbon tetrachloride and carbon disulfide.
ClC(Cl)Cl
119.37
CHEBI:35255
HPLC

Chemical Identifiers

67-66-3
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
ClC(Cl)Cl
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H331-H335-H336-H351-H361d-H373

DOTInformation : DOT Class: 6.1; Packing Group: III

EINECSNumber : 200-663-8

RTECSNumber : FS9100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only