Please login to your online account to display your discounted pricing

Chloroform, HPLC, 99.8% min., ACS, EMD Millipore

$278.89 - $278.89

Chemical Identifiers

Chemical Name or Material Chloroform
CAS 67-66-3
Name Note For ACS analysis
Purity Grade ACS/HPLC/NF/USP
Molecular Formula CHCl3
Linear Formula CHCl3
Formula Weight 119.38
Synonym Trichloromethane
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

MCX10501

 
emd millipore corporation
CX10501
Each for $278.89
Description & Specifications

Specifications

Chemical Name or Material Chloroform
Name Note For ACS analysis
CAS 67-66-3
Recommended Storage Room Temperature
Purity Grade ACS/HPLC/NF/USP
Physical Form Liquid
Assay Percent Range 99.8%, min.
Molecular Formula CHCl3
Linear Formula CHCl3
Formula Weight 119.38
Synonym Trichloromethane
Color Clear
Quantity 4L
Safety and Handling

Room Temperature