Please login to your online account to display your discounted pricing

Chloroform, For Spectroscopy Uvasol™, EMD Millipore

For spectroscopy Uvasol™

$119.43 - $449.49

Chemical Identifiers

Chemical Name or Material Chloroform
CAS 67-66-3
Molecular Formula CHCl3
Formula Weight 119.38
UN Number 1888
Synonym TCM, Trichloromethane, Methane trichloride
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1024470500

 
emd millipore corporation
1024470500
Glass Bottle 500mL Each for $119.43

M1024472500

 
emd millipore corporation
1024472500
Glass Bottle 2.5L Each for $449.49
Description & Specifications

Specifications

Melting Point −63°C
Formula Weight 119.38
Boiling Point 61°C
Quantity 500mL
Description The Uvasol™ solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The Uvasol™ solvents offer best UV transmittance.
Health Hazard 2 H315: Causes skin irritation.
CAS 67-66-3
Solubility Information Soluble in water: 8g/L at 20°C
Color Colorless
Packaging Glass Bottle
Odor Sweet
Density 1.4g/cm3 at 20°C
Chemical Name or Material Chloroform
Molecular Formula CHCl3
Vapor Pressure 211 hPa (20°C)
Storage Information 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition.
Synonym TCM, Trichloromethane, Methane trichloride
Physical Form Liquid
UN Number 1888
Health Hazard 1 H302: Harmful if swallowed.
Health Hazard 3 H319: Causes serious eye irritation.
Safety and Handling

H302: Harmful if swallowed.

H319: Causes serious eye irritation.

H315: Causes skin irritation.