Please login to your online account to display your discounted pricing

Chloroform (Ethanol as Preservative, Spectranalyzed™), Fisher Chemical


Quantity 4L
Packaging Glass Bottle

Manufacturer: fisher scientific company  BPC574SK4

 View more versions of this product

Catalog No. C574SK-4

  • / Each
  • / Case of 4 Each
 

Description & Specifications

Chemical Identifiers

Boiling Point 61°C
Formula Weight 119.38
Melting Point −63°C
Quantity 4L
DOT Information DOT Class 6.1, : Poison
Assay ≥99.8%
Vapor Pressure 213mbar at 20°C
Purity Grade Spectranalyzed™
Viscosity 0.56 mPa/s at 20°C
Decomposition Information 290°C
Physical Form Liquid
Color Colorless
Density 1.480g/cm³
Residue after Evaporation 2ppm max.
Assay Percent Range ≥99.8 %
Lead (Pb) 0.05ppm max.
MDL Number MFCD00000826
Concentration or Composition (by Analyte or Components) Approx. 0.75% Ethanol as preservative
Health Hazard 2 WARNING!
CAS 64-17-5
Health Hazard 3 Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:0
Acid and Chloride Pass Test
Identification Pass Test
Additional Information Vapor Pressure: 213mbar at 20°C
Packaging Glass Bottle
Substances Darkened by H2SO4 Pass Test
Infrared Spectrum Conforms to Ref
Acetone and Aldehyde Pass Test (about 0.005% Maximum)
Cleanup Symbol Solvent
Name Note Approx. 0.75% ethanol as preservative
Molecular Formula CHCl3
Absorbance Pass Test
ChemAlert Storage Symbol Blue
Synonym Trichloromethane
Free Chlorine Pass Test
Chemical Name or Material Chloroform
Purity Grade Notes Also meets ACS specifications
CAS 67-66-3

Specifications

Chemical Name or Material Chloroform
Name Note Approx. 0.75% ethanol as preservative
CAS 67-66-3
CAS 64-17-5
Purity Grade Spectranalyzed™
Purity Grade Notes Also meets ACS specifications
Assay ≥99.8%
Assay Percent Range ≥99.8 %
Molecular Formula CHCl3
Formula Weight 119.38
MDL Number MFCD00000826
Synonym Trichloromethane
Accessories
 

Emergency Overview
Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes and skin. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. May be harmful if inhaled. May cause irritation of respiratory tract. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:2
Flammability:1
Instability:0

WARNING!