Please login to your online account to display your discounted pricing

Chloroform, Environmental Grade, 99.8+%, stab. with ethanol, Alfa Aesar™

Suitable For Pesticide Analysis, Environmental and GC Analysis

$62.19 - $62.19

Chemical Identifiers

Chemical Name or Material Chloroform
CAS 67-66-3
Purity Grade Environmental Grade
Physical Form Liquid
Assay Percent Range >99.8%
Molecular Formula CHCl3
Formula Weight 119.38
MDL Number MFCD00000826
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40974K2

  SDS
alfa aesar
40974K2
1L Each for $62.19

AA40974K7

  SDS
alfa aesar
40974K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA40974M1

  SDS
alfa aesar
40974M1
4 x 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 119.38
Physical Form Liquid
Color Clear colorless
Quantity 1L
DOT Information DOT Class: 6.1, Packaging Group: III
Chemical Name or Material Chloroform
Ignition Point 982°C (1800°F)
EINECS Number 200-663-8
Odor Characteristic
Vapor Pressure 210 hPa (158mm Hg) at 20°C
Purity Grade Environmental Grade
Appearance Colorless liquid
Assay Percent Range >99.8%
Molecular Formula CHCl3
MDL Number MFCD00000826
CAS 67-66-3
Health Hazard 1 H351-H373-H302-H315: Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Causes skin irritation.
Safety and Handling

H351-H373-H302-H315: Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Causes skin irritation.