Please login to your online account to display your discounted pricing

Chloroform, EMD Millipore


Quantity 1L
Packaging Glass Bottle

Manufacturer: emd millipore corporation  CX10596

 View more versions of this product

Catalog No. MCX10596

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 119.38
Quantity 1L
CAS 67-66-3
Packaging Glass Bottle
Linear Formula CHCl3
Chemical Name or Material Chloroform

Specifications

Chemical Name or Material Chloroform
CAS 67-66-3
Linear Formula CHCl3
Formula Weight 119.38