missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform - D1, deuteration degree min. 99.96% for NMR spectroscopy, MagniSolv™, MilliporeSigma™

For NMR Spectroscopy

Manufacturer:  MilliporeSigma 1024460010

 View more versions of this product

Catalog No. M1024460010



Specifications

Specifications

Chloroform - D1
Glass ampoule
Colorless
865-49-6
CHCl3
HEDRZPFGACZZDS-MICDWDOJSA-N
trichloro(deuterio)methane
71583
120.38
Characteristic
211 hPa at 20°C (68°F)
10mL
99.96% min.
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
C(Cl)(Cl)Cl
120.375
CHEBI:85365
Liquid

Chemical Identifiers

865-49-6
120.375
chloroform-d, deuterochloroform, methane-d, trichloro, 2h chloroform, deuterated chloroform, cdcl3, trichloromethane-d, unii-p1nw4885vt, trichloro deuterio methane, chloroform, deutero
CHEBI:85365
C(Cl)(Cl)Cl
CHCl3
HEDRZPFGACZZDS-MICDWDOJSA-N
71583
trichloro(deuterio)methane
Documents
Promotions

Promotions