missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform - D1, deuteration degree min. 99.96% for NMR spectroscopy, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$63.48 - $348.71

Chemical Identifiers

CAS 865-49-6
Physical Form Liquid
Assay Percent Range 99.96% min.
Molecular Formula CDCl3
Formula Weight 120.38
Synonym Deuterochloroform
Chemical Name or Material Chloroform - D1
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
M1024460009 Emd Millipore Corporation
1024460009
Glass ampoule 10 x 0.75mL Each for $73.52
M1024460025 Emd Millipore Corporation
1024460025
Glass bottle 25mL Each for $115.11
M1024460010 Emd Millipore Corporation
1024460010
Glass ampoule 10mL Each for $63.48
M1024460100 Emd Millipore Corporation
1024460100
Glass bottle 100mL Each for $348.71
Specifications Description & Specifications

Chemical Identifiers

865-49-6
99.96% min.
120.38
Chloroform - D1
Liquid
CDCl3
Deuterochloroform

Specifications

865-49-6
Glass ampoule
Liquid
99.96% min.
CDCl3
Deuterochloroform
Characteristic
211 hPa at 20°C (68°F)
10 x 0.75mL
Colorless
120.38
Chloroform - D1