Please login to your online account to display your discounted pricing

Chloroform - D1, deuteration degree min. 99.8% for NMR spectroscopy, MagniSolv™, EMD Millipore™

For NMR Spectroscopy

$45.52 - $471.68

Chemical Identifiers

Chemical Name or Material Chloroform - D1
Name Note Stabilized with silver
CAS 865-49-6
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula CDCl3
Formula Weight 120.38
Synonym Deuterochloroform
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1024500100

 
emd millipore corporation
1024500100
Glass Bottle 100mL Each for $114.97

M1024500025

 
emd millipore corporation
1024500025
Glass bottle 25mL Each for $45.96

M1024500500

 
emd millipore corporation
1024500500
Glass bottle 500mL Each for $471.68

M1034200500

 
emd millipore corporation
1034200500
Glass bottle 500mL Each for $471.68

M1034200025

 
emd millipore corporation
1034200025
Glass bottle 25mL Each for $45.52

M1034200100

 
emd millipore corporation
1034200100
Glass bottle 100mL Each for $114.97
Description & Specifications

Specifications

Boiling Point 60°C
Melting Point −64.1°C
Formula Weight 120.38
Quantity 100mL
Color Colorless
Density 1.50g/cm3 at 20°C (68°F)
UN Number 1888
Packaging Glass Bottle
Physical Form Liquid
Chemical Name or Material Chloroform - D1
Vapor Pressure 211 hPa at 20°C (68°F)
Name Note Stabilized with silver
Assay Percent Range 99.8% min.
Molecular Formula CDCl3
CAS 865-49-6
Synonym Deuterochloroform
Odor Characteristic