Please login to your online account to display your discounted pricing

Chloroform - D1, 0.03 vol.% TMS, deuteration deg. min. 99.8% for NMR, MagniSolv™, EMD Millipore™

$54.33 - $561.07

Chemical Identifiers

Chemical Name or Material Chloroform - D1
Name Note 0.03 vol.% TMS, Stabilized with silver
CAS 865-49-6
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula CDCl3
Formula Weight 120.38
Synonym Deuterochloroform
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1032960025

 
emd millipore corporation
1032960025
Glass bottle 25mL Each for $54.33

M1032960100

 
emd millipore corporation
1032960100
Glass bottle 100mL Each for $137.35

M1032960500

 
emd millipore corporation
1032960500
Glass bottle 500mL Each for $561.07
Description & Specifications

Specifications

Formula Weight 120.38
Quantity 25mL
Color Colorless
Physical Form Liquid
Chemical Name or Material Chloroform - D1
Vapor Pressure 211 hPa at 20°C (68°F)
Name Note 0.03 vol.% TMS, Stabilized with silver
Assay Percent Range 99.8% min.
Molecular Formula CDCl3
CAS 865-49-6
Synonym Deuterochloroform
Odor Characteristic
Packaging Glass bottle