Please login to your online account to display your discounted pricing

Chloroform-d, for NMR, 99.8% atom D, stabilized with silver foil, ACROS Organics™

$46.54 - $511.61

Chemical Identifiers

Chemical Name or Material Chloroform-d
Name Note 99.8 atom% D
CAS 865-49-6
Purity Grade For NMR
Molecular Formula CCl3D
Linear Formula CDCl3
Formula Weight 120.39
MDL Number MFCD00000827
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC351420250

  SDS
thermo fisher scientific
351420250
25mL Each for $46.54

AC351421000

  SDS
thermo fisher scientific
351421000
100mL Each for $117.52

AC351425000

  SDS
thermo fisher scientific
351425000
500mL Each for $511.61
Description & Specifications

Specifications

Physical Form Liquid
Formula Weight 120.39
Quantity 25mL
Color Clear colorless
Beilstein 01, IV, 54
EINECS Number 212-742-4
Purity Grade For NMR
Chemical Name or Material Chloroform-d
Name Note 99.8 atom% D
Molecular Formula CCl3D
Linear Formula CDCl3
MDL Number MFCD00000827
CAS 865-49-6
Chlorine (Cl) 0.0001% max.
Deuterium (D) 99.75% min.
Water 0.02% max. (Coulometric)
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhale
Health Hazard 3 GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Ta
Packaging Glass bottle
Hydrochloric Acid 0.001% max.

For NMR

Safety and Handling

Warning

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Ta

GHS P Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhale