Please login to your online account to display your discounted pricing

Chloroform-d, for NMR, 99.8% atom D, packaged in 1mL ampoules, ACROS Organics™

$50.01 - $50.01

Chemical Identifiers

Chemical Name or Material Chloroform-d
Name Note 99.8 atom % D
CAS 865-49-6
Purity Grade For NMR
Molecular Formula CCl3D
Linear Formula CDCl3
Formula Weight 120.39
MDL Number MFCD00000827
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC217310100

  SDS
thermo fisher scientific
217310100
Each for $50.01
Description & Specifications

Specifications

Chemical Name or Material Chloroform-d
Name Note 99.8 atom % D
CAS 865-49-6
Deuterium (D) 99.75% min.
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness. Suspected of damaging the unborn child.
Health Hazard 3 GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
EINECS Number 212-742-4
Water 0.02% max. (Coulometric)
Purity Grade For NMR
Physical Form Liquid
Quantity 10 x 1mL
Color Clear colorless
Molecular Formula CCl3D
Linear Formula CDCl3
Formula Weight 120.39
MDL Number MFCD00000827
Beilstein 01, IV, 54

For NMR

Safety and Handling

Warning

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS H Statement
Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness. Suspected of damaging the unborn child.