Please login to your online account to display your discounted pricing

Chloroform-d, for NMR, 99.8% atom D, contains 1 v/v% TMS, ACROS Organics™

$27.27 - $173.32

Chemical Identifiers

Chemical Name or Material Chloroform-d
Name Note 99.8 atom % D
CAS 865-49-6
Purity Grade For NMR
Molecular Formula CCl3D
Linear Formula CDCl3
Formula Weight 120.39
MDL Number MFCD00000827
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166260500

  SDS
thermo fisher scientific
166260500
Glass bottle 50mL Each for $47.00

AC166261000

  SDS
thermo fisher scientific
166261000
Glass bottle 100mL Each for $79.82

AC166260250

  SDS
thermo fisher scientific
166260250
Glass bottle 25mL Each for $27.27

AC166262500

  SDS
thermo fisher scientific
166262500
Glass bottle 250mL Each for $173.32
Description & Specifications

Specifications

Formula Weight 120.39
Physical Form Crystals or Crystalline Powder
Quantity 50mL
Color White
Purity Grade For NMR
EINECS Number 212-742-4
Beilstein 01, IV, 54
Packaging Glass bottle
Chemical Name or Material Chloroform-d
Deuterium (D) 99.75% min.
CAS 865-49-6
MDL Number MFCD00000827
Linear Formula CDCl3
Molecular Formula CCl3D
Name Note 99.8 atom % D
Water 0.02% max. (Coulometric)
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Ta
Health Hazard 2 GHS P Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhale

For NMR

Safety and Handling

Warning

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Ta

GHS P Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhale