Please login to your online account to display your discounted pricing

Chloroform-d, for NMR, 99.8+% atom D, contains 0.03 v/v% TMS, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  209565000

 View more versions of this product

Catalog No. AC209565000

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 120.39
Physical Form Crystalline Powder
Quantity 500mL
Color White to beige
Beilstein 01, IV, 54
EINECS Number 212-742-4
Purity Grade For NMR
Water 0.01% max. (Coulometric)
Molecular Formula CCl3D
Packaging Glass bottle
Chemical Name or Material Chloroform-d
Name Note 99.8+ atom % D
Linear Formula CDCl3
MDL Number MFCD00000827
CAS 865-49-6
Deuterium (D) 99.80% min.
Health Hazard 1 Warning
Health Hazard 2 GHS P Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhale
Health Hazard 3 GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Ta

Specifications

Chemical Name or Material Chloroform-d
Name Note 99.8+ atom % D
CAS 865-49-6
Purity Grade For NMR
Molecular Formula CCl3D
Linear Formula CDCl3
Formula Weight 120.39
MDL Number MFCD00000827
Accessories

Warning

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Ta

GHS P Statement Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhale

SDS & Safety Documents