missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform-d, for NMR, 99.8+% atom D, contains 0.03 v/v% TMS, ACROS Organics™

$30.65 - $364.92

Chemical Identifiers

CAS 865-49-6
Molecular Formula CCl3D
Linear Formula CDCl3
Formula Weight 120.39
MDL Number MFCD00000827
Chemical Name or Material Chloroform-d, 99.8+ atom % D
Grade For NMR
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC209561000 SDS Thermo Fisher Scientific
209561000
Glass bottle 100mL Each for $88.94
AC209560250 SDS Thermo Fisher Scientific
209560250
Glass bottle 25mL Each for $30.65
AC209565000 SDS Thermo Fisher Scientific
209565000
Glass bottle 500mL Each for $364.92
Specifications

Description

For NMR
Description & Specifications

Chemical Identifiers

865-49-6
CDCl3
MFCD00000827
For NMR
CCl3D
120.39
Chloroform-d, 99.8+ atom % D

Specifications

99.80% min.
0.01% max. (Coulometric)
100mL
Crystalline Powder
CDCl3
MFCD00000827
Chloroform-d, 99.8+ atom % D
Glass bottle
White to beige
865-49-6
CCl3D
120.39
01, IV, 54
For NMR

Safety and Handling

GHS H Statement
Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness. Suspected of damaging the unborn child.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

EINECSNumber : 212-742-4