missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Chloroform-d, for NMR, 100.0% atom D, ACROS Organics™

$89.82 - $312.49

Chemical Identifiers

CAS 865-49-6
Molecular Formula CHCl3
Molecular Weight (g/mol) 120.375
MDL Number MFCD00000827
InChI Key HEDRZPFGACZZDS-MICDWDOJSA-N
Synonym 2h chloroform
PubChem CID 71583
ChEBI CHEBI:85365
IUPAC Name trichloro(deuterio)methane
SMILES C(Cl)(Cl)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC166270100 SDS View Documents Thermo Fisher Scientific
166270100
10mL Glass bottle Each for $89.82
 
AC166270500 SDS View Documents Thermo Fisher Scientific
166270500
50mL Glass bottle Each for $268.95
 
AC166271000 SDS View Documents Thermo Fisher Scientific
166271000
100mL Glass bottle Each for $312.49
 
Description

Description

For NMR
Specifications

Chemical Identifiers

865-49-6
120.375
HEDRZPFGACZZDS-MICDWDOJSA-N
71583
trichloro(deuterio)methane
CHCl3
MFCD00000827
2h chloroform
CHEBI:85365
C(Cl)(Cl)Cl

Specifications

99.95% min.
0.01% max. (Coulometric)
10mL
1.5000g/mL
-64.0°C
CDCl3
01, IV, 54
HEDRZPFGACZZDS-MICDWDOJSA-N
trichloro(deuterio)methane
71583
120.39
NMR
Glass bottle
Colorless
865-49-6
60.0°C
CHCl3
MFCD00000827
2h chloroform
C(Cl)(Cl)Cl
120.375
CHEBI:85365
Liquid
Chloroform-d, 100.0 atom % D
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness. Suspected of damaging the unborn child.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

EINECSNumber : 212-742-4